Learn More
Mettler Toledo™ RS232 to USB Converter Cable for XPR Microbalances and Ultra-Microbalances
Catalog No. 01911578
Change view
Click to view available options
Connection Type:
RS-232 and USB
Catalog No. Connection Type
01911578 RS-232 and USB
1 options
Catalog No. 01911578 Supplier Mettler Toledo™ Supplier No. 64088427
RS232 - USB converter cable to connect a balance (RS232) to a USB port
  • Cable with converter to connect a balance (RS232) to a USB port
  • For XPR Micro and Ultra-Microbalances
Specifications
For Use With (Equipment) XPR Micro and Ultra-Microbalance
Connection Type RS-232 and USB
For Use With (Application) Connects peripheral devices to the balance via RS2
Type USB-RS232 Converter