Learn More
OHAUS™ RS232 Interface Kit
Catalog No. 02112307
Change view
Click to view available options
RS232 Interface Kit
Catalog No.
02112307 RS232 Interface Kit
1 options
Catalog No. 02112307 Supplier OHAUS™ Supplier No. 30037448
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
RS232 PC board, pressure button, four screws, instruction manual
For use with Ranger 3000, Ranger Count 3000 compact counting scales and Valor 7000 scale.
For use with Ranger 3000, Ranger Count 3000 compact counting scales and Valor 7000 scale.
Specifications
For Use With (Equipment) Ranger 3000, Ranger Count 3000 Compact Counting Scales
Includes RS232 PC board, pressure button, four screws, inst
Type RS232 Interface Kit