missing translation for 'onlineSavingsMsg'
Learn More

Sartorius RS232 Interface Data Cable

RS232 Interface Data Cable for Entris™ Lab Balance.

$121.63 - $121.63

Specifications

For Use With (Equipment) Connection of a Secura, Quintix or Practum to computer, printer or second display.
Length (English) 5 ft.
Length (Metric) 1.5 m
Type Data Cable
View More Specs

Products 2
Catalog Number Mfr. No. Connection Type Price Quantity  
Catalog Number Mfr. No. Connection Type Price Quantity  
14558849
View Documents
Sartorius
YCCD09MUSBA
RS232 9-pin (male) to USB type A
Pack for $121.63
Only null left
 
14558851
View Documents
Sartorius
YCCD09MF
RS232 9-pin (male) to RS232 9-pin (female)
Pack for $121.63
Only null left
 
Description

Description

  • To connect Sartorius laboratory balances of the product family Entris™ to an external device via RS232 interface
  • Color: Black
Specifications

Specifications

Connection of a Secura, Quintix or Practum to computer, printer or second display.
5 ft.
1.5 m
Data Cable
Videos
SDS
Documents

Documents