missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(RS)-Mevalonic acid lithium salt, ≥96.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 150-97-0 |
| Physical Form | Neat |
| Linear Formula | C6H11O4 · xLi+ |
| Synonym | (+/-)-MVA-Li; rac.-MVA-Li; Lithium (+/-)-3,5-dihydroxy-3-methyl-pentanoate; Lithium (+/-)-3,5-dihydroxy-3-methyl-valerate |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 147.15 (free acid basis) |
| Percent Purity | ≥96.0% (GC) |
| Grade | Analytical Standard |