missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPRM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150096
Description
RPRM Polyclonal Antibody for Western Blot, ICC/IF, IHC
May be involved in the regulation of p53-dependent G2 arrest of the cell cycle. Seems to induce cell cycle arrest by inhibiting CDK1 activity and nuclear translocation of the CDC2 cyclin B1 complex.Specifications
| RPRM | |
| Polyclonal | |
| Unconjugated | |
| RPRM | |
| 2410012A13Rik; candidate mediator of the p53 dependent G2 arrest; candidate mediator of the p53-dependent G2 arrest; FLJ90327; protein reprimo; Reprimo; reprimo, TP53 dependant G2 arrest mediator candidate; reprimo, TP53 dependent G2 arrest mediator candidate; Rprm | |
| Rabbit | |
| Protein A | |
| RUO | |
| 56475, 67874, 680110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5BJN9, Q9JJ72, Q9NS64 | |
| RPRM | |
| Recombinant Reprimo (RPRM) (Met1-Tyr109). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |