Learn More
MP Biomedicals™ RPMI 1640 Medium, 1X Liq. without L-Glutamine, L-Cysteine, L-Cystine and L-Methionine
General purpose medium based on the RPMI 1630 line of media
Supplier: MP Biomedicals™ 091646454
Description
RPMI 1640 Medium was originally developed by Moore and his colleagues at Roswell Park Memorial Institute (RPMI). It was based on the RPMI 1630 line of media which utillized a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins.
RPMI has successfully been used for the cultivation of normal human and neoplastic leukocytes. It is now a popular general purpose medium when properly supplemented.
Specifications
| Liquid | |
| Sterile | |
| 500 mL | |
| 4°C |
| RPMI 1640 | |
| No Glutamine, No Cystine, No Cysteine, No Methionine | |
| 6.8 to 7.4 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.