missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPL32 Recombinant Rabbit Monoclonal Antibody (HL2331)
GREENER_CHOICE

Catalog No. FEMA550492
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550492 Supplier Invitrogen™ Supplier No. MA550492

Rabbit Recombinant Monoclonal Antibody

RPL32 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC, IHC (P)

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L32E family of ribosomal proteins. It is located in the cytoplasm. Although some studies have mapped this gene to 3q13. 3-q21, it is believed to map to 3p25-p24. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternatively spliced transcript variants encoding the same protein have been observed for this gene.

Specifications

Antigen RPL32
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2331
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Rpl32
Gene Accession No. E2RKA8, P62910, P62911, P62912
Gene Alias 60S ribosomal protein L32; AU020185; hypothetical protein LOC615556; hypothetical protein LOC677748; L32; Large ribosomal subunit protein eL32; PP9932; ribosomal protein L32; RPL32; rpL32-3A; snoRNA MBI-141; zgc:136952
Gene Symbols Rpl32
Host Species Rabbit
Immunogen Full length of human RPL32 recombinant protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19951, 28298, 556556, 607481, 6161
Target Species Human, Mouse, Rat, Canine, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less