missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL15 Polyclonal Antibody

Description
RPL15 Polyclonal Antibody for Western Blot, ELISA
May play a role as co-chaperone of the Hsp70 family proteins HSPA1A, HSPA1B and HSPA8.
Specifications
Specifications
| Antigen | RPL15 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | Rpl15 |
| Gene Accession No. | P61313 |
| Gene Alias | 2510008H07Rik; 60S ribosomal protein L15; DBA12; EC45; L15; Large ribosomal subunit protein eL15; ribosomal protein L15; RPL10; RPL15; RPLY10; RPYL10; TCBAP0781 |
| Gene Symbols | Rpl15 |
| Show More |