missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPL11 Monoclonal Antibody (3A4A7)
GREENER_CHOICE

Catalog No. 373000
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
373000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 373000 Supplier Invitrogen™ Supplier No. 373000
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

RPL11 Monoclonal Antibody for ELISA, ICC/IF, Western Blot

Subunit of the splicing factor SF3B required for 'A' complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA. May also be involved in the assembly of the 'E' complex. Belongs also to the minor U12-dependent spliceosome, which is involved in the splicing of rare class of nuclear pre-mRNA intron.

Specifications

Antigen RPL11
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3A4A7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Rpl11
Gene Accession No. P62913
Gene Alias 2010203J19Rik; 60S ribosomal protein L11; cell growth-inhibiting protein 34; CLL-associated antigen KW-12; DBA7; GIG34; L11; Large ribosomal subunit protein uL5; ribosomal protein L11; RP11-223J15.3; RPL11; zgc:86748
Gene Symbols Rpl11
Host Species Mouse
Immunogen fusing the mouse myeloma cell-line P3U1 with spleen cells of BALB/c mouse after immunization with human Laminin (1.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6135
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less