missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPA70 Recombinant Rabbit Monoclonal Antibody (23GB4885)
GREENER_CHOICE

Catalog No. MA553828
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553828 100 μL
MA553829 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553828 Supplier Invitrogen™ Supplier No. MA553828
Only null left

Rabbit Recombinant Monoclonal Antibody

RPA70 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene Plays an essential role in several cellular processes in DNA metabolism including replication, recombination and DNA repair. Binds and subsequently stabilizes single-stranded DNA intermediates and thus prevents complementary DNA from reannealing.

Specifications

Antigen RPA70
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4885
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPA1
Gene Accession No. P27694
Gene Alias 5031405K23Rik; 70kDa; AA589576; AW557552; Cb1-727; HSSB; MST075; MSTP075; REPA1; Replication factor A protein 1; replication protein A 70 kDa DNA-binding subunit; Replication protein A 70 kDa DNA-binding subunit, N-terminally processed; replication protein A1; replication protein A1, 70kDa; RF-A; RF-A protein 1; Rpa; RP-A; RP-A p70; RPA1; RPA70; Single-stranded DNA-binding protein
Gene Symbols RPA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPA70 (1-54AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287524, 6117
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less