missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18612609
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612609 50 kg Poly Bottle
18612604 25 g Poly Bottle
18612605 125 g Poly Bottle
18612606 500 g Poly Bottle
18612607 2.5 kg Poly Bottle
18612608 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18612609 Supplier Spectrum Chemical Mfg Cor Supplier No. P128250KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7778-50-9

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 50 kg
Molecular Weight (g/mol) 294.18
Percent Purity 99%
Grade ACS/Reagent
Loss on Drying 0.0005
Packaging Poly Bottle
Chemical Name or Material Potassium Dichromate, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.