missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Roxithromycin 95.0+%, TCI America™
$105.89 - $284.92
Chemical Identifiers
| CAS | 80214-83-1 |
|---|---|
| Molecular Formula | C41H76N2O15 |
| Molecular Weight (g/mol) | 837.06 |
| MDL Number | MFCD00214389 |
| InChI Key | RXZBMPWDPOLZGW-KMAKEOJNSA-N |
| Synonym | Erythromycin 9-[O-(2-Methoxyethoxy)methyloxime] |
| PubChem CID | 133687104 |
| IUPAC Name | (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-6-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-[(5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl)oxy]-3,5,7,9,11,13-hexamethyl-10-(2,4,7-trioxa-1-azaoctan-1-ylidene)-1-oxacyclotetradecan-2-one |
| SMILES | CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2CC(C)(OC)C(O)C(C)O2)[C@H](C)[C@@H](OC2OC(C)CC(C2O)N(C)C)[C@](C)(O)C[C@@H](C)\C(=N/OCOCCOC)[C@H](C)[C@@H](O)[C@]1(C)O |
Chemical Identifiers
| 80214-83-1 | |
| 837.06 | |
| RXZBMPWDPOLZGW-KMAKEOJNSA-N | |
| 133687104 | |
| CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2CC(C)(OC)C(O)C(C)O2)[C@H](C)[C@@H](OC2OC(C)CC(C2O)N(C)C)[C@](C)(O)C[C@@H](C)\C(=N/OCOCCOC)[C@H](C)[C@@H](O)[C@]1(C)O |
| C41H76N2O15 | |
| MFCD00214389 | |
| Erythromycin 9-[O-(2-Methoxyethoxy)methyloxime] | |
| (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-6-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-[(5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl)oxy]-3,5,7,9,11,13-hexamethyl-10-(2,4,7-trioxa-1-azaoctan-1-ylidene)-1-oxacyclotetradecan-2-one |
Specifications
| Roxithromycin | |
| 80214-83-1 | |
| White | |
| C41H76N2O15 | |
| Erythromycin 9-[O-(2-Methoxyethoxy)methyloxime] | |
| CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2CC(C)(OC)C(O)C(C)O2)[C@H](C)[C@@H](OC2OC(C)CC(C2O)N(C)C)[C@](C)(O)C[C@@H](C)\C(=N/OCOCCOC)[C@H](C)[C@@H](O)[C@]1(C)O | |
| 837.06 | |
| 837.06 | |
| Roxithromycin |
| Crystalline Powder | |
| 120°C | |
| 5 g | |
| MFCD00214389 | |
| RXZBMPWDPOLZGW-KMAKEOJNSA-N | |
| (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-6-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-[(5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl)oxy]-3,5,7,9,11,13-hexamethyl-10-(2,4,7-trioxa-1-azaoctan-1-ylidene)-1-oxacyclotetradecan-2-one | |
| 133687104 | |
| ≥95.0% (HPLC) |
Safety and Handling
RTECSNumber : KF4990000
TSCA : No
Recommended Storage : Refrigerator