missing translation for 'onlineSavingsMsg'
Learn More

Roxatidine Acetate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135743
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
R0178200MG 200 mg
R01781G 1 g
2 options

Catalog No. R0178200MG

Supplier: TCI America R0178200MG

Only null left

Chemical Identifiers

CAS 93793-83-0
Molecular Formula C19H29ClN2O4
Molecular Weight (g/mol) 384.901
MDL Number MFCD00941429
InChI Key FEWCTJHCXOHWNL-UHFFFAOYSA-N
Synonym 2-Acetoxy-N-[3-[3-(1-piperidinylmethyl)phenoxy]propyl]acetamide Hydrochloride
PubChem CID 56704
IUPAC Name [2-oxo-2-[3-[3-(piperidin-1-ylmethyl)phenoxy]propylamino]ethyl] acetate;hydrochloride
SMILES CC(=O)OCC(=O)NCCCOC1=CC=CC(=C1)CN2CCCCC2.Cl

Specifications

Melting Point 148°C
Color White
Quantity 200 mg
Formula Weight 384.90
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Roxatidine Acetate Hydrochloride
RTECSNumber AC3855000
TSCA No