accessibility menu, dialog, popup

UserName

CAS RN 93793-83-0

IUPAC Name: hydrogen [(3-{3-[(piperidin-1-yl)methyl]phenoxy}propyl)carbamoyl]methyl acetate chloride
Synonyms: hydrogen roxatidine acetate chloride
Molecular Weight (g/mol): 384.9
Molecular Formula: C19H29ClN2O4
InChi Key: FEWCTJHCXOHWNL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(=O)OCC(=O)NCCCOC1=CC=CC(CN2CCCCC2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results