missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Round Bottom Polypropylene Test Tubes for Storage Box
For use with Micro Rack Tube System.
Supplier: Greiner Bio-One 102270
Description
Polypropylene Test Tubes display high thermal, mechanical and chemical resistance. It is therefore recommended for the storage of chemical and biological samples
- Volume: 1.2mL
- Dia. x H: 8.5 x 44mm
- Available with or without
Specifications
| 8-Tube chain | |
| 44 mm | |
| Micro Rack Tube System; Storage of chemical and biological samples | |
| 8.5 mm | |
| Test Tube |
| 1.2 mL | |
| Polypropylene | |
| Non-sterile | |
| 8-Strip Chain |