missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Inc Rotenone, ≥97%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215015410

 View more versions of this product

Catalog No. ICN15015410



Description

Description

Specifications

Specifications

Rotenone
Beige to White
C23H22O6
rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root
CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC
6758
394.4
83-79-4
≥97%
10 g
JUVIOZPCNVVQFO-HBGVWJBISA-N
394.423
CHEBI:28201
Powder

Chemical Identifiers

83-79-4
394.423
rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root
CHEBI:28201
C23H22O6
JUVIOZPCNVVQFO-HBGVWJBISA-N
6758
CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC
SDS
Documents

Documents

Promotions

Promotions