missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rotavirus NCDV Polyclonal Antibody, Biotin

Goat Polyclonal Antibody

Supplier:  Invitrogen™ PA173016

Catalog No. PIPA173016


Only null left
Explore available promotions

Description

Description

The PA1-73016 antibody reacts with Rotavirus Bovine NCDV. PA1-73016 has been successfully used in ELISA applications. The PA1-73016 antibody was raised against Rotavirus ICP + virions. Recommended short term storage of less than 6 months store at 4°C. For extended storage, store at -20°C.

Rotavirus is a double-stranded RNA virus in the family Reoviridae. Five species of rotavirus (A-E) cause disease in animals with species A being the most common in human. Within rotavirus A there are different strains, called serotypes, based on the dual classification system of two proteins on the surface of the virus. Six viral proteins (VPs), or structural proteins, form the virion and six non-structural proteins (NSPs) are expressed in infected cells. Rotavirus is the most common cause of severe diarrhoea among infants and young children. Rotavirus gastroenteritis is a mild to severe disease characterised by vomiting, watery diarrhoea, and low-grade fever, but still leads to hundreds of thousands of deaths each year in developing countries.
Specifications

Specifications

Rotavirus NCDV
Polyclonal
Biotin
Reoviridae
Bovine (Nebraska Calf Diarrhea Virus).
1 mL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Goat
IgG fraction
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions