missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rosmarinic Acid, 98.6%, MP Biomedicals™
$74.22 - $74.22
Chemical Identifiers
| CAS | 20283-92-5 |
|---|---|
| Molecular Formula | C18H16O8 |
| Molecular Weight (g/mol) | 360.318 |
| MDL Number | MFCD00017740 |
| InChI Key | DOUMFZQKYFQNTF-WUTVXBCWSA-N |
| Synonym | rosmarinic acid, rosemary acid, r-rosmarinic acid, unii-mqe6xg29yi, rosmarinate, mqe6xg29yi, labiatenic acid, r-o-3,4-dihydroxycinnamoyl-3-3,4-dihydroxyphenyl lactic acid, 3,4-dihydroxycinnamic acid r-1-carboxy-2-3,4-dihydroxyphenyl ethyl ester, 2r-3-3,4-dihydroxyphenyl-2-2e-3-3,4-dihydroxyphenyl prop-2-enoyloxy propanoic acid |
| PubChem CID | 5281792 |
| ChEBI | CHEBI:50371 |
| IUPAC Name | (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid |
| SMILES | C1=CC(=C(C=C1CC(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15979210
|
MP Biomedicals Inc
0215979210 |
10 mg |
Each for $74.22
|
|
|||||
Chemical Identifiers
| 20283-92-5 | |
| 360.318 | |
| DOUMFZQKYFQNTF-WUTVXBCWSA-N | |
| 5281792 | |
| (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid |
| C18H16O8 | |
| MFCD00017740 | |
| rosmarinic acid, rosemary acid, r-rosmarinic acid, unii-mqe6xg29yi, rosmarinate, mqe6xg29yi, labiatenic acid, r-o-3,4-dihydroxycinnamoyl-3-3,4-dihydroxyphenyl lactic acid, 3,4-dihydroxycinnamic acid r-1-carboxy-2-3,4-dihydroxyphenyl ethyl ester, 2r-3-3,4-dihydroxyphenyl-2-2e-3-3,4-dihydroxyphenyl prop-2-enoyloxy propanoic acid | |
| CHEBI:50371 | |
| C1=CC(=C(C=C1CC(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)O)O |
Specifications
| 20283-92-5 | |
| 0.986 | |
| MFCD00017740 | |
| rosmarinic acid, rosemary acid, r-rosmarinic acid, unii-mqe6xg29yi, rosmarinate, mqe6xg29yi, labiatenic acid, r-o-3,4-dihydroxycinnamoyl-3-3,4-dihydroxyphenyl lactic acid, 3,4-dihydroxycinnamic acid r-1-carboxy-2-3,4-dihydroxyphenyl ethyl ester, 2r-3-3,4-dihydroxyphenyl-2-2e-3-3,4-dihydroxyphenyl prop-2-enoyloxy propanoic acid | |
| C1=CC(=C(C=C1CC(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)O)O | |
| 360.318 | |
| CHEBI:50371 | |
| Powder |
| Beige | |
| C18H16O8 | |
| 10 mg | |
| DOUMFZQKYFQNTF-WUTVXBCWSA-N | |
| (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid | |
| 5281792 | |
| 360.32 | |
| Rosmarinic acid, Molecular Biology |
Safety and Handling
Recommended Storage : Store at room temperature, protect from light.