missing translation for 'onlineSavingsMsg'
Learn More

Rosmarinic Acid, 98.6%, MP Biomedicals™

Catalog No. ICN15979210
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15979210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15979210 Supplier MP Biomedicals Inc Supplier No. 0215979210

Chemical Identifiers

CAS 20283-92-5
Molecular Formula C18H16O8
Molecular Weight (g/mol) 360.318
MDL Number MFCD00017740
InChI Key DOUMFZQKYFQNTF-WUTVXBCWSA-N
Synonym rosmarinic acid, rosemary acid, r-rosmarinic acid, unii-mqe6xg29yi, rosmarinate, mqe6xg29yi, labiatenic acid, r-o-3,4-dihydroxycinnamoyl-3-3,4-dihydroxyphenyl lactic acid, 3,4-dihydroxycinnamic acid r-1-carboxy-2-3,4-dihydroxyphenyl ethyl ester, 2r-3-3,4-dihydroxyphenyl-2-2e-3-3,4-dihydroxyphenyl prop-2-enoyloxy propanoic acid
PubChem CID 5281792
ChEBI CHEBI:50371
IUPAC Name (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid
SMILES C1=CC(=C(C=C1CC(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)O)O

Specifications

Color Beige
Assay Percent Range 0.986
Quantity 10 mg
Formula Weight 360.32
Physical Form Powder
Chemical Name or Material Rosmarinic acid, Molecular Biology
Recommended Storage Store at room temperature, protect from light.