missing translation for 'onlineSavingsMsg'
Learn More

(-)-Rose oxide, ≥99% (Mixture of Isomers (cis+trans), GC), MilliporeSigma™ Supelco™

Catalog No. 111013298
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013298 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013298 Supplier Merck Emd Millipore Supplier No. 839171ML
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 16409-43-1
Density 0.873 g/mL (at 20°C)
Flash Point 69°C
Physical Form Neat
Linear Formula C10H18 O
MDL Number MFCD00036607
Synonym (4 R)-2-(2-Methyl-1-propenyl)-4-methyltetrahydropyran
Type Analytical Standard
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥99% (mixture of isomers (cis+trans), GC)
Grade Analytical Standard
Show More Show Less