missing translation for 'onlineSavingsMsg'
Learn More

Rose Bengal, Spectrum™ Chemical

$378.50 - $4,733.29

Chemical Identifiers

CAS 632-69-9
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18613041
View Documents Promotion Details
N/A 10 g Amber Glass Bottle
Each for $378.50
Only null left
 
18613042
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $551.68
Only null left
 
18613043
View Documents Promotion Details
N/A 500 g Amber Glass Bottle
Each for $4,733.29
Only null left
 
Description

Description

Specifications

Chemical Identifiers

632-69-9

Specifications

632-69-9
Reagent
Rose Bengal
1
Brown to Red
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.