missing translation for 'onlineSavingsMsg'
Learn More

Rose Bengal, Spectrum™ Chemical

Catalog No. 18613043
Change view
Click to view available options
Quantity:
10 g
25 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613043 500 g Amber Glass Bottle
18613041 10 g Amber Glass Bottle
18613042 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613043 Supplier Spectrum Chemical Mfg Cor Supplier No. RO115500GMBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 632-69-9

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Brown to Red
Quantity 500 g
Molecular Weight (g/mol) 1017.64
Grade Reagent
Chemical Name or Material Rose Bengal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.