Learn More
Promega ROS-Glo™ H2O2 Assay
The ROS-Glo™ H2O2 Assay is a homogeneous, fast and sensitive bioluminescent assay that measures the level of hydrogen peroxide (H2O2), a reactive oxygen species (ROS), directly in cell culture or in defined enzyme reactions.
$863.03 - $3,607.59
Specifications
Content And Storage -30°C to -10°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRG8820
Encompass
View Documents Promotion Details
Promega
G8820
10 mL
Each for $863.03
Only null left
PRG8821
Encompass
View Documents Promotion Details
Promega
G8821
50 mL
Each for $3,607.59
Only null left
Description
Description
  • Specific, Direct Detection of Hydrogen Peroxide (H2O2)
  • Fast assay involves a simple two-reagent-addition protocol
  • Substrate reacts directly with H2O2; no need for HRP as a coupling enzyme
Specifications
Specifications
-30°C to -10°C