missing translation for 'onlineSavingsMsg'
Learn More

Promega ROS-Glo™ H2O2 Assay

Catalog No. PRG8821
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG8821 50 mL
PRG8820 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG8821 Supplier Promega Supplier No. G8821

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The ROS-Glo™ H2O2 Assay is a homogeneous, fast and sensitive bioluminescent assay that measures the level of hydrogen peroxide (H2O2), a reactive oxygen species (ROS), directly in cell culture or in defined enzyme reactions.

  • Specific, Direct Detection of Hydrogen Peroxide (H2O2)
  • Fast assay involves a simple two-reagent-addition protocol
  • Substrate reacts directly with H2O2; no need for HRP as a coupling enzyme

Specifications

Quantity 50 mL
Content And Storage -30°C to -10°C