missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ ROR gamma (t) [del. 5-8] Plasmid
Description
The ROR gamma (t) [del. 5-8] Plasmid is a mammalian expression vector construct, pCMV/hRORgamma (t) [del. 5-8], containing coding sequence of a novel isoform of human RAR-related orphan receptor gamma (t) which lacks the hingeencoding exons 5 - 8 and represses IL-17A gene transcription1.
- Species: Human
- Gene: RORC
- Immunogen: ROR gamma (t) [del. 5-8] Plasmid
- Total contents of 10μg is enough for one 96-well cell transfection
Recommended Storage
Store at -20°C. Avoid freeze-thaw cycles.
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Product Type | ROR gamma (t) [del. 5-8] Plasmid |
| Quantity | 10 μg |