missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Roll Form Pipe Markers, Legend: OXYGEN

Ideal for identifying pipe content and flow without the hassle of precise measurement around the pipe's diameter. Brady Roll Form Pipe Marker, Legend: OXYGEN, is made of pressure sensitive vinyl

$113.46

Specifications

Material Pressure-sensitive Vinyl
Quantity 1 Roll
Size (English) 30 ft.x 8 in.
Size (Metric) 9.14 m x 20.3 cm
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19112073
View Documents Promotion Details
Brady™
41469
Black, Yellow
Each for $113.46
Only null left
 
19112074
View Documents Promotion Details
Brady™
41470
White, Blue
Each for $113.46
Only null left
 
Description

Description

  • Wrap around entire width of pipe and adhere to themselves
  • Legend repeats 65 times per roll
  • For marking pipes that carry same contents
  • With vinyl adhesive suited for use in outdoor applications
Specifications

Specifications

Pressure-sensitive Vinyl
1 Roll
30 ft.x 8 in.
9.14 m x 20.3 cm
Videos
SDS
Documents

Documents