missing translation for 'onlineSavingsMsg'
Learn More

Rolipram 98.0+%, TCI America™

Catalog No. R011050MG
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R011050MG 50 mg
R0110250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R011050MG Supplier TCI America Supplier No. R011050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61413-54-5
Molecular Formula C16H21NO3
Molecular Weight (g/mol) 275.35
MDL Number MFCD00270906
InChI Key HJORMJIFDVBMOB-UHFFFAOYNA-N
Synonym rolipram, rolipramum, rolipramum latin, 4-3-cyclopentyloxy-4-methoxyphenyl-2-pyrrolidinone, adeo, +/--rolipram, 4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, rolipram usan:inn, 3h rolipram
PubChem CID 5092
ChEBI CHEBI:104872
IUPAC Name 4-[3-(cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
SMILES COC1=CC=C(C=C1OC1CCCC1)C1CNC(=O)C1

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 50 mg
Formula Weight 275.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rolipram
RTECSNumber UY5749237
TSCA No
Recommended Storage Freezer (-20 °C)