missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rolipram, 98%

Catalog No. AAJ67177MA
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67177MA 10 mg
AAJ67177LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67177MA Supplier Thermo Scientific Chemicals Supplier No. J67177MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61413-54-5 | C16H21NO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61413-54-5
Molecular Formula C16H21NO3
Synonym 4-(3-Cyclopentyloxy-4-methoxyphenyl)-2-pyrrolidinone; ZK 62711

Specifications

Quantity 10 mg
Description White to off-white
Formula Weight 275.35g/mol
Form Solid
Assay 98%

RUO – Research Use Only