missing translation for 'onlineSavingsMsg'
Learn More

Ro60 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501565803
Encompass
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501565803 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501565803 Supplier Bethyl Laboratories Inc Supplier No. A303693AT
Only null left

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.

Specifications

Antigen Ro60
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TROVE2
Gene Accession No. O08848, P10155
Gene Alias RP11-101E13.3, RO60, RORNP, SSA2, 60 kDa SS-A/Ro ribonucleoprotein, 60 kDa ribonucleoprotein Ro, Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro), gastric cancer multi-drug resistance protein, sjoegren syndrome type A antigen
Gene Symbols TROVE2
Host Species Rabbit
Immunogen Between 488 and 538
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20822, 6738
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less