missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF186 Polyclonal Antibody

Catalog No. PIPA542330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542330 Supplier Invitrogen™ Supplier No. PA542330

Rabbit Polyclonal Antibody

Peptide sequence: MACTKTLQQS QPISAGATTT TTAVAPAGGH SGSTECDLEC LVCREPYSCP Sequence homology: Human: 100%.

The RNF186 gene encodes the RING finger protein 186, a member of the RING-type E3 ubiquitin ligase family. RNF186 is involved in the ubiquitin-proteasome pathway, playing a crucial role in tagging target proteins for degradation, thus influencing protein turnover and cellular homeostasis. This process is essential for regulating various cellular functions, including signal transduction, apoptosis, and response to stress. RNF186 has been implicated in the unfolded protein response (UPR) within the endoplasmic reticulum, where it helps in managing protein folding and maintaining cellular homeostasis, particularly under conditions of endoplasmic reticulum stress. Alterations in RNF186 expression or function have been associated with disorders such as inflammatory bowel disease and ulcerative colitis, suggesting its role in gastrointestinal health and disease. Research into RNF186 focuses on its regulatory mechanisms within the ubiquitin-proteasome system and its potential as a therapeutic target for conditions associated with protein misfolding and cellular stress responses.

Specifications

Antigen RNF186
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF186
Gene Accession No. Q9NXI6
Gene Alias 9130020G10Rik; E3 ubiquitin-protein ligase RNF186; RING finger protein 186; RNF186
Gene Symbols RNF186
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RNF186 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54546
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less