missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF146 Monoclonal Antibody (K16280_8D7), Invitrogen™
Description
Sequence of this protein is as follows: NLARESSADG ADSVSAQSGA SVQPLVSSVR PLTSVDGQLT SPATPSPDAS TSLEDSFAHL QLSGDNTAER SHRGEGEEDH ESPSSGRVPA PDTSIEETES DASSDSEDVS AVVAQHSLTQ QRLLVSNANQ
RNF146 protein function is currently unknown.
Specifications
Specifications
| Antigen | RNF146 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | K16280_8D7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | RNF146 |
| Gene Accession No. | Q5XIK5, Q9NTX7 |
| Gene Alias | 2610509H23Rik; dactylidin; E3 ubiquitin-protein ligase RNF146; iduna; RING finger protein 146; RING finger protein 146; E3 ubiquitin-protein ligase RNF146; RING-type E3 ubiquitin transferase RNF146; Rnf146 |
| Show More |