missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF146 Monoclonal Antibody (K16280_8D7), Invitrogen™

Catalog No. MA555652
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555652 50 μL
MA556032 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555652 Supplier Invitrogen™ Supplier No. MA555652
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: NLARESSADG ADSVSAQSGA SVQPLVSSVR PLTSVDGQLT SPATPSPDAS TSLEDSFAHL QLSGDNTAER SHRGEGEEDH ESPSSGRVPA PDTSIEETES DASSDSEDVS AVVAQHSLTQ QRLLVSNANQ

RNF146 protein function is currently unknown.

Specifications

Antigen RNF146
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K16280_8D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene RNF146
Gene Accession No. Q5XIK5, Q9NTX7
Gene Alias 2610509H23Rik; dactylidin; E3 ubiquitin-protein ligase RNF146; iduna; RING finger protein 146; RING finger protein 146; E3 ubiquitin-protein ligase RNF146; RING-type E3 ubiquitin transferase RNF146; Rnf146
Gene Symbols RNF146
Host Species Mouse
Immunogen Recombinant human RNF146 (immunogen range is 191-320).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308051, 81847
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less