missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF144A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101573
Description
Antibody detects endogenous levels of total RNF144A.
UBCE7IP4 (Ubiquitin-conjugating enzyme 7- interacting protein 4), also known as RNF144A (RING finger protein 144A), KIAA0161 or RNF144, is a 292 amino acid single-pass membrane protein that contains one RING-type zinc finger and two IBR-type zinc fingers. Functioning as an E3 ubiquitin-protein ligase, UBCE7IP4 accepts ubiquitin (in the form of a thioester) from E2 ubiquitin-conjugating enzymes, such as UBC8 and UBCH7, and transfers that ubiquitin residue to target substrates. Via its RING finger, UBCE7IP4 may play a role in protein-DNA and protein-protein interactions throughout the cell.Chemical Identifiers
| RNF144A | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| RNF144A | |
| E3 ubiquitin-protein ligase RNF144A; Kiaa0161; probable E3 ubiquitin-protein ligase RNF144A; RGD1561737; ring finger protein 144; ring finger protein 144A; RNF144; RNF144A; Ubce7ip4; UbcM4-interacting protein 4; ubiquitin conjugating enzyme 7 interacting protein 4; ubiquitin-conjugating enzyme 7-interacting protein 4; Uip4 | |
| RNF144A | |
| Primary | |
| Affinity chromatography |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P50876, Q925F3 | |
| 108089, 9781 | |
| A synthesized peptide derived from human RNF144A(Accession P50876), corresponding to amino acid residues H151-M201. | |
| Antibody |
Specifications
| RNF144A | |
| Polyclonal | |
| Unconjugated | |
| RNF144A | |
| E3 ubiquitin-protein ligase RNF144A; Kiaa0161; probable E3 ubiquitin-protein ligase RNF144A; RGD1561737; ring finger protein 144; ring finger protein 144A; RNF144; RNF144A; Ubce7ip4; UbcM4-interacting protein 4; ubiquitin conjugating enzyme 7 interacting protein 4; ubiquitin-conjugating enzyme 7-interacting protein 4; Uip4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108089, 9781 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P50876, Q925F3 | |
| RNF144A | |
| A synthesized peptide derived from human RNF144A(Accession P50876), corresponding to amino acid residues H151-M201. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |