missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF130 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553181
Description
Immunogen sequence: PWLSEHCTCP MCKLNILKAL GIVPNLPCTD NVAFDMERLT RTQAVNRRSA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.
The protein encoded by this gene contains a RING finger motif and is similar to g1, a Drosophila zinc-finger protein that is expressed in mesoderm and involved in embryonic development. The expression of the mouse counterpart was found to be upregulated in myeloblastic cells following IL3 deprivation, suggesting that this gene may regulate growth factor withdrawal-induced apoptosis of myeloid precursor cells. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2013].Chemical Identifiers
| RNF130 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| RNF130 | |
| 2510042A13Rik; E3 ubiquitin-protein ligase RNF130; g1-related zinc finger protein; G1rp; G1RZFP; GOLIATH; Goliath homolog; GP; H-Goliath; R-goliath; ring finger protein 130; RING-type E3 ubiquitin transferase RNF130; RNF130; rnf130 {ECO:0000250; UniProtKB:Q86XS8} | |
| RNF130 | |
| Primary | |
| Antigen affinity chromatography |
| 0.1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86XS8 | |
| 55819 | |
| Recombinant protein corresponding to Human RNF130. Recombinant protein control fragment (Product #RP-90935). | |
| Antibody |
Specifications
| RNF130 | |
| Polyclonal | |
| Unconjugated | |
| RNF130 | |
| 2510042A13Rik; E3 ubiquitin-protein ligase RNF130; g1-related zinc finger protein; G1rp; G1RZFP; GOLIATH; Goliath homolog; GP; H-Goliath; R-goliath; ring finger protein 130; RING-type E3 ubiquitin transferase RNF130; RNF130; rnf130 {ECO:0000250; UniProtKB:Q86XS8} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55819 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86XS8 | |
| RNF130 | |
| Recombinant protein corresponding to Human RNF130. Recombinant protein control fragment (Product #RP-90935). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |