missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF113B Monoclonal Antibody (OTI2C10)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525822
Description
RNF113B Monoclonal Antibody for Western Blot, IHC (P), Flow
RNF113B (Ring Finger Protein 113B) is a Protein Coding gene. An important paralog of this gene is RNF113A.Chemical Identifiers
| RNF113B | |
| 0.57 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IZP6 | |
| 140432 | |
| Full length human recombinant protein of RNF113B produced in HEK293T cell. | |
| Antibody |
| OTI2C10 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse, Rat, Canine, Monkey | |
| RNF113B | |
| bA10G5.1; RING finger protein 113B; RNF113B; RNF161; Zinc finger protein 183-like 1; ZNF183L1 | |
| RNF113B | |
| Primary | |
| Affinity Chromatography |
Specifications
| RNF113B | |
| Monoclonal | |
| 0.57 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IZP6 | |
| RNF113B | |
| Full length human recombinant protein of RNF113B produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C10 | |
| Unconjugated | |
| RNF113B | |
| bA10G5.1; RING finger protein 113B; RNF113B; RNF161; Zinc finger protein 183-like 1; ZNF183L1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 140432 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |