missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNASE8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5150270
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5150270 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5150270 Supplier Invitrogen™ Supplier No. PA5150270
Only null left

Rabbit Polyclonal Antibody

RNASE8 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

RNASE8 has a low ribonuclease activity.

Specifications

Antigen RNASE8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene RNASE8
Gene Accession No. Q8TDE3
Gene Alias RAE2; ribonuclease 8; ribonuclease A family member 8; ribonuclease, RNase A family, 8; RNase 8; RNase 8, placental; RNASE8
Gene Symbols RNASE8
Host Species Rabbit
Immunogen Recombinant Ribonuclease A8 (RNASE8) (Trp19-Val154).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122665
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less