missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNASE3 Monoclonal Antibody (C3)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA565968

Catalog No. MA565968

Only null left
Explore available promotions

Description

Description

RNASE3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

ECP (eosinophil cationic protein), also known as RNS3 or RNASE3 (ribonuclease, RNase A family, 3), is a 160 amino acid potent cytotoxic secretory protein that belongs to the pancreatic ribonuclease family. ECP localizes to the granule matrix of the eosinophil and is involved in the immune response system. ECP is a cytotoxin, neurotoxin and helminthotoxin that is secreted by activated human eosinophils. ECP exhibits anti-parasitic, antibacterial and ribonucleolytic activities. It has been suggested that ECP induces the neurotoxic effect known as the Gordon phenomenon, a syndrome manifested by ataxia, muscular rigidity, paralysis and tremor that may lead to death. ECP is considered a marker of eosinophilic inflammation and high levels have been found in cases of active asthma and other allergic diseases.
Specifications

Specifications

RNASE3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P12724
RNASE3
Recombinant Ribonuclease A3 (RNASE3) (Pro30-Val154).
200 μL
Primary
Human
Antibody
IgG2b λ
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
C3
Unconjugated
RNASE3
cytotoxic ribonuclease; Ear11; EAR-11; Ear3; ECP; Eosinophil cationic protein; eosinophil cationic protein-like; eosinophil cationic-type ribonuclease 3; Eosinophil secondary granule ribonuclease 11; eosinophil-associated ribonuclease 3; eosinophil-associated, ribonuclease A family, member 3; LOC100361866; mR3; MR-3; murine ribonuclease 3; RAF1; Ribonuclease 3; ribonuclease A family member 3; ribonuclease, RNase A family 3; ribonuclease, RNase A family, 3; RNase 3; RNASE3; RNS3
Mouse
Protein A/G
RUO
6037
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions