missing translation for 'onlineSavingsMsg'
Learn More

RNASE/DNASE-Free Water, MP Biomedicals™, 500mL

Catalog No. MP112450204
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP112450204 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112450204 Supplier MP Biomedicals Inc Supplier No. 112450204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Formula Weight 18.02
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Water, Not DEPC-treated.