missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAqueous™ Total RNA Isolation Kit

Catalog No. AM1912
Encompass
Change view
Click to view available options
Quantity:
50 Catridges
50 Preps
Product Type:
RNA Isolation Kit
Spin Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
AM1912 50 Preps RNA Isolation Kit
AM10051G 50 Catridges Spin Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM1912 Supplier Invitrogen™ Supplier No. AM1912
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 10^2–10^7 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

The RNAqueous Total RNA Isolation Kit is for phenol-free total RNA isolation from 1–75 mg of tissue or from 102–107 cells, using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

The RNAqueous kit provides:

  • Convenient syringe-based filtration
  • Compatibility with vacuum manifolds when processing multiple samples

How it works

The RNAqueous kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.

Specifications

Content And Storage • 6 mL Formaldehyde Load Dye; store at -20°C
• 60 mL Lysis/Binding Solution; 4°C
• 60 mL Water; 4°C
• 4 mL Lithium Chloride Precipitation Solution; 4°C
• 40 mL Wash Solution #1; 4°C
• 80 mL Wash Solution #2/3 (concentrated); 4°C
• 50 Filter Cartridges; room temperature
• 100 Collection Tubes; room temperature
• 10 mL Elution Solution; room temperature
Isolation Technology Spin Column (Glass Fiber Filter)
Elution Volume 50–120 μL
Sample Type Bacteria, Cells, Plant, Tissue, Yeast, Viruses, Enzymatic Reactions
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product Type RNA Isolation Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Starting Material Amount Bacteria: ≤109 cells, Cells: ≤107, Tissue: ≤75 mg, Plant: ≤75 mg, Yeast: ≤108 cells, Virus: varies, Enzymatic Reactions: varies
Show More Show Less
What is the minimum RNA size that is isolated using RNaqueous kits?

RNAs less than 200 nt are lost using the columns in the RNAqeous family of kits. These kits are not suitable for isolation of 5S rRNA or tRNA. The RNAqueous-Micro Total RNA Isolation Kit does, however, offer a modified protocol to allow isolation of miRNA from the sample.

I'm using your RNAqueous Total RNA Isolation Kit , and the filter is clogging. Do you have any suggestions?

To prevent filter clogging with viscous samples or samples containing a large amount of tissue, perform a clarifying centrifugatoin at 10,000-15,000 x g and remove to a fresh tube prior to adding ethanol to the lysis/binding solution.

What are the differences in the RNAqueous kits you offer?

RNaqueous kits are based on a rapid, filter-based RNA isolation system that does not require phenol, chloroform, or other toxic organic chemicals. The RNAqueous Total RNA Isolation Kit is designed for sample sizes of 10-75 mg of tissue or 10^6 to 10^7 cells, whereas the RNAqueous-Midi Total RNA Isolation Kit is designed for tissue samples of up to 0.5 g or 10^9 cells. The RNAqueous-Micro Total RNA Isolation Kit is optimized for the purification of total RNA from micro-sized samples such as 10-500,000 cultured cells, as few as 10 microdissected cells, or up to 10 mg tissue. The RNAqueous-96 Total RNA Isolation Kit utilizes a 96-well plate format for high-throughput RNA isolation from 100 to 2 x 10^6 cells or 0.1 to 1.5 mg of tissue. The RNAqueous-4PCR Total RNA Isolation Kit is designed for RNA isolation from small tissue samples (0.5-75 mg) and small- to medium-sized cultured cell samples (approximately 100-10^7 cells).

For Research Use Only. Not for use in diagnostic procedures.