missing translation for 'onlineSavingsMsg'
Learn More

Rizatriptan Benzoate 98.0+%, TCI America™

Catalog No. p-7135745
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R018150MG 50 mg
R0181250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R018150MG Supplier TCI America Supplier No. R018150MG
Only null left

Chemical Identifiers

CAS 145202-66-0
Molecular Formula C22H25N5O2
Molecular Weight (g/mol) 391.475
MDL Number MFCD00866224
InChI Key JPRXYLQNJJVCMZ-UHFFFAOYSA-N
Synonym rizatriptan benzoate, maxalt, maxalt-mlt, unii-wr978s7qhh, rizatrimptan benzoate, rizatriptane benzoate, rizatriptan benzoate usan:usp, n,n-dimethyl-2-5-1,2,4-triazol-1-ylmethyl-1h-indol-3-yl ethylamine benzoate, 2-5-1h-1,2,4-triazol-1-yl methyl-1h-indol-3-yl-n,n-dimethylethanamine benzoate
PubChem CID 77997
IUPAC Name benzoic acid;N,N-dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine
SMILES CN(C)CCC1=CNC2=C1C=C(C=C2)CN3C=NC=N3.C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 180°C
Color White
Quantity 50 mg
Formula Weight 391.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Rizatriptan Benzoate