accessibility menu, dialog, popup

UserName

CAS RN 145202-66-0

IUPAC Name: benzoic acid; dimethyl(2-{5-[(1H-1,2,4-triazol-1-yl)methyl]-1H-indol-3-yl}ethyl)amine
Synonyms: rizatriptan
Molecular Weight (g/mol): 391.48
Molecular Formula: C22H25N5O2
InChi Key: JPRXYLQNJJVCMZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1.CN(C)CCC1=CNC2=CC=C(CN3C=NC=N3)C=C12
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)