missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rizatriptan benzoate

Catalog No. AC462940010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462940010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462940010 Supplier Thermo Scientific Chemicals Supplier No. 462940010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145202-66-0
Molecular Formula C22H25N5O2
Molecular Weight (g/mol) 391.48
InChI Key JPRXYLQNJJVCMZ-UHFFFAOYSA-N
IUPAC Name benzoic acid; dimethyl(2-{5-[(1H-1,2,4-triazol-1-yl)methyl]-1H-indol-3-yl}ethyl)amine
SMILES OC(=O)C1=CC=CC=C1.CN(C)CCC1=CNC2=CC=C(CN3C=NC=N3)C=C12

Specifications

Melting Point 178.0°C to 180.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: 53 g/L Other solubilities: soluble in methanol, slightly soluble in ethanol, insoluble in ether
Formula Weight 391.47
Chemical Name or Material Rizatriptan benzoate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes serious eye irritation.

Precautionary Statement GHS P Statement Wear eye protection/face protection.

Wash face, hands and any exposed skin thoroughly after handling.

If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only