missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine Related Compound B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013710
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013710 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013710 Supplier Merck Emd Millipore Supplier No. PHR203030MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 25081-93-0
Linear Formula C13H20N2O2 · HCl
Synonym (RS)-3-[1-(Dimethylamino)ethyl]phenyl dimethylcarbamate hydrochloride
Type Certified Reference Material
Quantity 30 mg
Formula Weight 272.77
Grade Certified Reference Material