missing translation for 'onlineSavingsMsg'
Learn More

Rivastigmine Hydrogen Tartrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012912
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012912 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012912 Supplier Merck Emd Millipore Supplier No. PHR18671G

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 129101-54-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C14H22N2O2 · C4H6O6
Synonym Rivastigmine tartrate; ENA-713; Ethylmethyl-carbamic acid 3-[(1 S)-1-(dimethylamino)ethyl]phenyl ester; N-Ethyl-N-methyl-carbamic acid 3-[(1 S)-1-(dimethylamino)ethyl]phenyl ester tartrate; S-Rivastigmine tartrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 400.42
Grade Certified Reference Material