missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ritonavir, 98%, Thermo Scientific Chemicals
$253.73 - $253.73
Chemical Identifiers
| CAS | 155213-67-5 |
|---|---|
| Molecular Formula | C37H48N6O5S2 |
| Molecular Weight (g/mol) | 720.94 |
| InChI Key | NCDNCNXCDXHOMX-XGKFQTDJSA-N |
| Synonym | ritonavir, norvir, abbott 84538, abbott-84538, ritonavir usan, norvir softgel, norvir sec, unii-o3j8g9o825, norvir tm, norvir tn |
| PubChem CID | 392622 |
| ChEBI | CHEBI:45409 |
| IUPAC Name | 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate |
| SMILES | CC(C)C1=NC(=CS1)CN(C)C(=O)NC(C(C)C)C(=O)NC(CC2=CC=CC=C2)CC(C(CC3=CC=CC=C3)NC(=O)OCC4=CN=CS4)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC461222500
|
Thermo Scientific Chemicals
461222500 |
250 mg |
Each for $253.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 155213-67-5 | |
| 720.94 | |
| ritonavir, norvir, abbott 84538, abbott-84538, ritonavir usan, norvir softgel, norvir sec, unii-o3j8g9o825, norvir tm, norvir tn | |
| CHEBI:45409 | |
| CC(C)C1=NC(=CS1)CN(C)C(=O)NC(C(C)C)C(=O)NC(CC2=CC=CC=C2)CC(C(CC3=CC=CC=C3)NC(=O)OCC4=CN=CS4)O |
| C37H48N6O5S2 | |
| NCDNCNXCDXHOMX-XGKFQTDJSA-N | |
| 392622 | |
| 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate |
Specifications
| 155213-67-5 | |
| Authentic | |
| C37H48N6O5S2 | |
| ritonavir, norvir, abbott 84538, abbott-84538, ritonavir usan, norvir softgel, norvir sec, unii-o3j8g9o825, norvir tm, norvir tn | |
| NCDNCNXCDXHOMX-XGKFQTDJSA-N | |
| 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate | |
| 392622 | |
| 720.94 | |
| Gel |
| 120.0°C to 132.0°C | |
| 97.5% min. (HPLC) | |
| 250 mg | |
| Solubility in water: <1 mg/mL. Other solubilities: 20 mg/mL dmso, 1 mg/mL ethanol, slightly soluble in methanol, soluble in isopropanol | |
| CC(C)C1=NC(=CS1)CN(C)C(=O)NC(C(C)C)C(=O)NC(CC2=CC=CC=C2)CC(C(CC3=CC=CC=C3)NC(=O)OCC4=CN=CS4)O | |
| 720.94 | |
| CHEBI:45409 | |
| 98% | |
| Ritonavir |
Safety and Handling
Harmful if swallowed, in contact with skin or if inhaled
Acute toxicity (category 4)
RUO – Research Use Only