missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ritodrine Hydrochloride 98.0+%, TCI America™
$46.20 - $136.69
Chemical Identifiers
| CAS | 23239-51-2 |
|---|---|
| Molecular Formula | C17H22ClNO3 |
| Molecular Weight (g/mol) | 323.82 |
| MDL Number | MFCD01657514 |
| InChI Key | IDLSITKDRVDKRV-XHXSRVRCSA-N |
| Synonym | N-(4-Hydroxyphenethyl)-4-hydroxynorephedrine Hydrochloride |
| PubChem CID | 46911860 |
| IUPAC Name | hydrogen 4-(2-{[(1R,2S)-1-hydroxy-1-(4-hydroxyphenyl)propan-2-yl]amino}ethyl)phenol chloride |
| SMILES | [H+].[Cl-].C[C@H](NCCC1=CC=C(O)C=C1)[C@H](O)C1=CC=C(O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0114250MG
|
TCI America
R0114250MG |
250 mg |
Each for $46.20
|
|
|||||
|
R01141G
|
TCI America
R01141G |
1 g |
Each for $136.69
|
|
|||||
Chemical Identifiers
| 23239-51-2 | |
| 323.82 | |
| IDLSITKDRVDKRV-XHXSRVRCSA-N | |
| 46911860 | |
| [H+].[Cl-].C[C@H](NCCC1=CC=C(O)C=C1)[C@H](O)C1=CC=C(O)C=C1 |
| C17H22ClNO3 | |
| MFCD01657514 | |
| N-(4-Hydroxyphenethyl)-4-hydroxynorephedrine Hydrochloride | |
| hydrogen 4-(2-{[(1R,2S)-1-hydroxy-1-(4-hydroxyphenyl)propan-2-yl]amino}ethyl)phenol chloride |
Specifications
| 23239-51-2 | |
| White-Yellow | |
| MFCD01657514 | |
| N-(4-Hydroxyphenethyl)-4-hydroxynorephedrine Hydrochloride | |
| [H+].[Cl-].C[C@H](NCCC1=CC=C(O)C=C1)[C@H](O)C1=CC=C(O)C=C1 | |
| 323.82 | |
| 323.82 | |
| Crystalline Powder |
| 195°C | |
| C17H22ClNO3 | |
| 250 mg | |
| IDLSITKDRVDKRV-XHXSRVRCSA-N | |
| hydrogen 4-(2-{[(1R,2S)-1-hydroxy-1-(4-hydroxyphenyl)propan-2-yl]amino}ethyl)phenol chloride | |
| 46911860 | |
| ≥98.0% (HPLC,N) | |
| Ritodrine Hydrochloride |
Safety and Handling
RTECSNumber : DO6524000
TSCA : No