missing translation for 'onlineSavingsMsg'
Learn More

Ritodrine Hydrochloride 98.0+%, TCI America™

Catalog No. R01141G
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01141G 1 g
R0114250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01141G Supplier TCI America Supplier No. R01141G
Only null left

Chemical Identifiers

CAS 23239-51-2
Molecular Formula C17H22ClNO3
Molecular Weight (g/mol) 323.82
MDL Number MFCD01657514
InChI Key IDLSITKDRVDKRV-XHXSRVRCSA-N
Synonym N-(4-Hydroxyphenethyl)-4-hydroxynorephedrine Hydrochloride
PubChem CID 46911860
IUPAC Name hydrogen 4-(2-{[(1R,2S)-1-hydroxy-1-(4-hydroxyphenyl)propan-2-yl]amino}ethyl)phenol chloride
SMILES [H+].[Cl-].C[C@H](NCCC1=CC=C(O)C=C1)[C@H](O)C1=CC=C(O)C=C1

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 1 g
Formula Weight 323.82
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ritodrine Hydrochloride
RTECSNumber DO6524000
TSCA No