missing translation for 'onlineSavingsMsg'
Learn More

Ritodrine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013612
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013612 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013612 Supplier Merck Emd Millipore Supplier No. PHR16151G

Ritodrine Hydrochloride is a potent tocolytic drug and a selective γ2-adrenoceptor agonist, widely used for the control of premature labor.

Specifications

CAS 23239-51-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H21 NO3 · HCl
MDL Number MFCD01657514
Synonym Ritodrine hydrochloride; N-(p-Hydroxyphenethyl)-4-hydroxynorephedrine hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 323.81
Grade Certified Reference Material