Learn More
Thermo Scientific™ Ritalinic acid
Supplier: Thermo Scientific™ 463000050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Ritalinic acid | |
| C13H17NO2 | |
| INGSNVSERUZOAK-UHFFFAOYNA-N | |
| 2-phenyl-2-(piperidin-2-yl)acetic acid | |
| 219.28 |
| 19395-41-6 | |
| 5 g | |
| OC(=O)C(C1CCCCN1)C1=CC=CC=C1 | |
| 219.28 |
Chemical Identifiers
| 19395-41-6 | |
| 219.28 | |
| 2-phenyl-2-(piperidin-2-yl)acetic acid |
| C13H17NO2 | |
| INGSNVSERUZOAK-UHFFFAOYNA-N | |
| OC(=O)C(C1CCCCN1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement Causes serious eye irritation.
GHS P Statement Wear eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 243-020-7
RUO â Research Use Only