Learn More
Invitrogen™ Human RIPK5, GST Tag Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ PV4678
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
RIPK5 is a serine/threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis. This gene encodes a dual serine/threonine and tyrosine protein kinase which is expressed in multiple tissues. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| Q6XUX3 | |
| Kinase Assay | |
| 25778 | |
| Human RIPK5, GST Tag | |
| For research use only. Not for use in diagnostic procedures | |
| RIPK5 | |
| RIPK5 Recombinant Human Protein | |
| Human | |
| GST-tag | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Kinase and inhibitors | |
| Serine/threonine kinase |
| See Label | |
| 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 | |
| 132.7 kDa | |
| 10 μg | |
| A930019K20Rik; C430014H23Rik; C820013G01; CAKUT1; Dstyk; Dual serine/threonine and tyrosine protein kinase; dusty Pk.; Dusty protein kinase; DustyPK; HDCMD38P; KIAA0472; receptor interacting protein kinase 5; receptor-interacting serine/threonine-protein kinase 5; RIP5; RIP-homologous kinase; Ripk5; sgK496; Sugen kinase 496 | |
| RIPK | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Insect cells | |
| Liquid |